Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHRNB3 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297039DL594
Description
CHRNB3 Polyclonal specifically detects CHRNB3 in Human samples. It is validated for ELISA.Specifications
CHRNB3 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
DyLight 594 | |
acetylcholine receptor, neuronal nicotinic, beta-3 subunit, cholinergic receptor, nicotinic, beta 3, cholinergic receptor, nicotinic, beta polypeptide 3, neuronal acetylcholine receptor subunit beta-3 | |
Produced in rabbits immunized with purified, recombinant CHRNB3 (NP_000740.1; Met1-Leu232). | |
0.1 mL | |
Protein Phosphatase | |
1142 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction