Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRNB3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP17995020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17995020 20 μL
NBP179950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17995020 Supplier Novus Biologicals Supplier No. NBP17995020UL

Rabbit Polyclonal Antibody

CHRNB3 Polyclonal specifically detects CHRNB3 in Human samples. It is validated for Western Blot.

Specifications

Antigen CHRNB3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_000740
Gene Alias acetylcholine receptor, neuronal nicotinic, beta-3 subunit, cholinergic receptor, nicotinic, beta 3, cholinergic receptor, nicotinic, beta polypeptide 3, neuronal acetylcholine receptor subunit beta-3
Gene Symbols CHRNB3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CHRNB3The immunogen for this antibody is CHRNB3. Peptide sequence YDGTMVDLILINENVDRKDFFDNGEWEILNAKGMKGNRRDGVYSYPFITY.
Molecular Weight of Antigen 53 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1142
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.