Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHRNB3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572191
Description
CHRNB3 Polyclonal Antibody for Western Blot
The nicotinic acetylcholine receptors are members of a superfamily of ligand-gated ion channels that mediate fast signal transmission at synapses. The nAChRs are stated that the proposed structure for each subunit is a conserved N-terminal extracellular domain followed by 3 conserved transmembrane domains, a variable cytoplasmic loop, a fourth conserved transmembrane domain, and a short C-terminal extracellular region.
Specifications
CHRNB3 | |
Polyclonal | |
Unconjugated | |
Chrnb3 | |
5730417K16Rik; acetylcholine receptor; acetylcholine receptor, neuronal nicotinic, beta-3 subunit; acetylcholine receptor, nicotinic, beta 3 (neuronal); Acrb3; cholinergic receptor; cholinergic receptor nicotinic beta 3 subunit; cholinergic receptor, nicotinic beta 3; cholinergic receptor, nicotinic, beta 3 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 3; Chrnb3; I79_003985; Neuronal acetylcholine receptor subunit beta-3; neuronal nicotinic | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
100774746, 1142 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.34 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q05901 | |
Chrnb3 | |
KLH conjugated synthetic peptide between 187-200 amino acids from the Central region of human CHRNB3. | |
400 μL | |
Primary | |
Human, Hamster | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction