Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRNB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587636
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587636 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587636 Supplier Invitrogen™ Supplier No. PA587636
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IAENEDALLR HLFQGYQKWV RPVLHSNDTI KVYFGLKISQ LVDVDEKNQL MTTNVWLKQE WTDHKLRWNP DDYGGIHSIK VPSESLWLPD IVLFENADGR FEGSLMTKVI VKSNGTVVWT PPASYKSSCT MDVTFFPFDR QNCSMKFGSW TYDGTMVDLI LINENVDRKD FFDNGEWEIL NAKGMKGNRR DGVYSYPFIT YSFVLRRL; Positive Samples: Mouse brain, Mouse eye, Mouse liver, Mouse skeletal muscle, Rat liver; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse.

The nicotinic acetylcholine receptors are members of a superfamily of ligand-gated ion channels that mediate fast signal transmission at synapses. The nAChRs are stated that the proposed structure for each subunit is a conserved N-terminal extracellular domain followed by 3 conserved transmembrane domains, a variable cytoplasmic loop, a fourth conserved transmembrane domain, and a short C-terminal extracellular region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNB3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Chrnb3
Gene Accession No. P12391, Q05901, Q8BMN3
Gene Alias 5730417K16Rik; acetylcholine receptor; acetylcholine receptor, neuronal nicotinic, beta-3 subunit; acetylcholine receptor, nicotinic, beta 3 (neuronal); Acrb3; cholinergic receptor; cholinergic receptor nicotinic beta 3 subunit; cholinergic receptor, nicotinic beta 3; cholinergic receptor, nicotinic, beta 3 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 3; Chrnb3; I79_003985; Neuronal acetylcholine receptor subunit beta-3; neuronal nicotinic
Gene Symbols Chrnb3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 25-232 of human CHRNB3 (NP_0007401).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108043, 1142, 171131
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.