Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHRNB4 Monoclonal Antibody (7F5B2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531694
Description
MA5-31694 has been tested in indirect ELISA.
After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
Specifications
CHRNB4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P30926 | |
CHRNB4 | |
Purified recombinant fragment of human CHRNB4 (AA: extra 22-236) expressed in E. coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
7F5B2 | |
Unconjugated | |
CHRNB4 | |
acetylcholine receptor; Acetylcholine receptor beta 4; acetylcholine receptor, nicotinic, beta 4 (neuronal); Acrb4; Acrb-4; cholinergic receptor; cholinergic receptor nicotinic beta 4 subunit; cholinergic receptor, nicotinic beta 4; cholinergic receptor, nicotinic, beta 4 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 4; Chrnb4; nAChRB4; N-alpha 2; neuronal acetylcholine receptor non-alpha-2 chain; Neuronal acetylcholine receptor subunit beta-4; neuronal nicotinic receptor beta 4 subunit; nicotinic acetylcholine receptor beta4 subunit | |
Mouse | |
Protein G | |
RUO | |
1143 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction