Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRNB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142356
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142356 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142356 Supplier Invitrogen™ Supplier No. PA5142356
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: The immunizing peptide represents part of the first extracellular domain (aa22-236). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.

After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNB4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CHRNB4
Gene Accession No. P30926
Gene Alias acetylcholine receptor; Acetylcholine receptor beta 4; acetylcholine receptor, nicotinic, beta 4 (neuronal); Acrb4; Acrb-4; cholinergic receptor; cholinergic receptor nicotinic beta 4 subunit; cholinergic receptor, nicotinic beta 4; cholinergic receptor, nicotinic, beta 4 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 4; Chrnb4; nAChRB4; N-alpha 2; neuronal acetylcholine receptor non-alpha-2 chain; Neuronal acetylcholine receptor subunit beta-4; neuronal nicotinic receptor beta 4 subunit; nicotinic acetylcholine receptor beta4 subunit
Gene Symbols CHRNB4
Host Species Goat
Immunogen Peptide with sequence C-DYRLTWNSSRYEGVN.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1143
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.