Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRND Mouse anti-Human, Clone:1H1F9, Abnova™

Catalog No. 89334323 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-323 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-323 Supplier Abnova Corporation Supplier No. MAB17970
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human CHRND.

The acetylcholine receptor of muscle has 5 subunits of 4 different types: 2 alpha and 1 each of beta, gamma and delta subunits. After acetylcholine binding, the receptor undergoes an extensive conformation change that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane. [provided by RefSeq]

Specifications

Antigen CHRND
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1H1F9
Conjugate Unconjugated
Description cholinergic receptor, nicotinic, delta
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene CHRND
Gene Alias ACHRD, CMS2A, FCCMS, SCCMS
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 22-245 of human CHRND.
Quantity 100 μg
Gene ID (Entrez) 1144
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.