Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRND Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587924 Supplier Invitrogen™ Supplier No. PA587924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNEEERLIRH LFQEKGYNKE LRPVAHKEES VDVALALTLS NLISLKEVEE TLTTNVWIEH GWTDNRLKWN AEEFGNISVL RLPPDMVWLP EIVLENNNDG SFQISYSCNV LVYHYGFVYW LPPAIFRSSC PISVTYFPFD WQNCSLKFSS LKYTAKEITL SLKQDAKENR TYPVEWIIID PEGFTENGEW EIVHRPARVN VDPRAPLDSP SRQDITFYLI IRRK; Positive Samples: A-431, 293T, HeLa, Mouse lung; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse.

The acetylcholine receptor of muscle has 5 subunits of 4 different types: 2 alpha and 1 each of beta, gamma and delta subunits. After acetylcholine binding, the receptor undergoes an extensive conformation change that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRND
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CHRND
Gene Accession No. P02716, Q07001
Gene Alias acetylcholine receptor; acetylcholine receptor delta subunit; acetylcholine receptor subunit delta; acetylcholine receptor, nicotinic, delta (muscle); AChR delta subunit; Achr-4; ACHRD; Acrd; cholinergic receptor; cholinergic receptor nicotinic delta subunit; cholinergic receptor, nicotinic delta; cholinergic receptor, nicotinic, delta (muscle); cholinergic receptor, nicotinic, delta polypeptide; CHRND; CMS2A; CMS3A; CMS3B; CMS3C; delta subunit of muscle acetylcholine receptor; FCCMS; L10076; nicotinic acetylcholine receptor delta polypeptide; SCCMS; sofa potato; sop; wu:fa01e01; zgc:110469
Gene Symbols CHRND
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 22-245 of human CHRND (NP_0007421).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1144, 11447
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.