Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRND Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981950
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1950 Supplier Bioss Supplier No. BS12114R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The acetylcholine receptor of muscle has 5 subunits of 4 different types: 2 alpha and 1 each of beta, gamma and delta subunits. After acetylcholine binding, the receptor undergoes an extensive conformation change that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.

Specifications

Antigen CHRND
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CHRND
Gene Accession No. Q07001
Gene Alias acetylcholine receptor delta subunit; Acetylcholine receptor subunit delta; acetylcholine receptor, nicotinic, delta (muscle); AChR delta subunit; Achr-4; ACHRD; Acrd; BOS_2155; cholinergic receptor nicotinic delta subunit; cholinergic receptor, nicotinic delta; cholinergic receptor, nicotinic, delta; cholinergic receptor, nicotinic, delta (muscle); cholinergic receptor, nicotinic, delta polypeptide; CHRND; CMS2A; CMS3A; CMS3B; CMS3C; FCCMS; L10076; nicotinic acetylcholine receptor delta polypeptide; SCCMS; sofa potato; sop; wu:fa01e01; zgc:110469
Gene Symbols CHRND
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CHRND.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1144
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.