Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRNE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587600
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587600 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587600 Supplier Invitrogen™ Supplier No. PA587600
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KNEELRLYHH LFNNYDPGSR PVREPEDTVT ISLKVTLTNL ISLNEKEETL TTSVWIGIDW QDYRLNYSKD DFGGIETLRV PSELVWLPEI VLENNIDGQF GVAYDANVLV YEGGSVTWLP PAIYRSVCAV EVTYFPFDWQ NCSLIFRSQT YNAEEVEFTF AVDNDGKTIN KIDIDTEAYT ENGEWAIDFC PGVIRRHHGG ATDGPGETDV IYSLIIRRK; Positive Samples: HepG2, Mouse skeletal muscle, Rat kidney, Rat skeletal muscle; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse.

After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNE
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CHRNE
Gene Accession No. P09660, P20782, Q04844
Gene Alias acetylcholine receptor; acetylcholine receptor epsilon; acetylcholine receptor subunit epsilon; acetylcholine receptor, nicotinic, epsilon (muscle); AchR epsilon subunit; Achre; AChrepsilon; ACHRG; Acre; cholinergic receptor; cholinergic receptor nicotinic epsilon subunit; cholinergic receptor, nicotinic epsilon; cholinergic receptor, nicotinic, epsilon (muscle); cholinergic receptor, nicotinic, epsilon polypeptide; CHRNE; CMS1D; CMS1E; CMS2A; CMS4A; CMS4B; CMS4C; epsilon-subunit; FCCMS; gene for acetylcholine receptor epsilon subunit; MGC133376; nAChRE; nicotinic acetylcholine receptor epsilon subunit; SCCMS
Gene Symbols CHRNE
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 21-239 of human CHRNE (NP_0000711).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11448, 1145, 29422
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.