Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRNG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568078 Supplier Invitrogen™ Supplier No. PA568078
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

The mammalian muscle-type acetylcholine receptor is a transmembrane pentameric glycoprotein with two alpha subunits, one beta, one delta, and one epsilon (in adult skeletal muscle) or gamma (in fetal and denervated muscle) subunit. This gene, which encodes the gamma subunit, is expressed prior to the thirty-third week of gestation in humans. The gamma subunit of the acetylcholine receptor plays a role in neuromuscular organogenesis and ligand binding and disruption of gamma subunit expression prevents the correct localization of the receptor in cell membranes. Mutations in this gene cause Escobar syndrome and a lethal form of multiple pterygium syndrome. Muscle-type acetylcholine receptor is the major antigen in the autoimmune disease myasthenia gravis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNG
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHRNG
Gene Accession No. P04760, P07510, P18916
Gene Alias acetylcholine receptor; Acetylcholine receptor subunit gamma; acetylcholine receptor, muscle, gamma subunit; acetylcholine receptor, nicotinic, gamma (muscle); AChR gamma subunit; Achr-3; ACHRG; Acrg; cholinergic receptor; cholinergic receptor nicotinic gamma subunit; cholinergic receptor, nicotinic gamma; cholinergic receptor, nicotinic, gamma (muscle); cholinergic receptor, nicotinic, gamma polypeptide; Chrng; chrng variant N1; chrng variant N2; nicotinic acetylcholine receptor gamma subunit
Gene Symbols CHRNG
Host Species Rabbit
Immunogen Synthesized peptide derived from human Acetylcholine Receptor gamma.
Purification Method affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11449, 1146, 25753
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.