Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHRNG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531170
Description
Recommended positive controls: 293T, A431. Predicted reactivity: Mouse (82%), Rat (82%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The mammalian muscle-type acetylcholine receptor is a transmembrane pentameric glycoprotein with two alpha subunits, one beta, one delta, and one epsilon (in adult skeletal muscle) or gamma (in fetal and denervated muscle) subunit. This gene, which encodes the gamma subunit, is expressed prior to the thirty-third week of gestation in humans. The gamma subunit of the acetylcholine receptor plays a role in neuromuscular organogenesis and ligand binding and disruption of gamma subunit expression prevents the correct localization of the receptor in cell membranes. Mutations in this gene cause Escobar syndrome and a lethal form of multiple pterygium syndrome. Muscle-type acetylcholine receptor is the major antigen in the autoimmune disease myasthenia gravis.
Specifications
CHRNG | |
Polyclonal | |
Unconjugated | |
CHRNG | |
acetylcholine receptor; Acetylcholine receptor subunit gamma; acetylcholine receptor, muscle, gamma subunit; acetylcholine receptor, nicotinic, gamma (muscle); AChR gamma subunit; Achr-3; ACHRG; Acrg; cholinergic receptor; cholinergic receptor nicotinic gamma subunit; cholinergic receptor, nicotinic gamma; cholinergic receptor, nicotinic, gamma (muscle); cholinergic receptor, nicotinic, gamma polypeptide; Chrng; chrng variant N1; chrng variant N2; nicotinic acetylcholine receptor gamma subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1146 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P07510 | |
CHRNG | |
Recombinant protein encompassing a sequence within the center region of human AChR gamma. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction