Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Chroma-Glo™ Luciferase Assay System

Catalog No. pre4920
Click to view available options
Quantity:
100 mL

The Chroma-Glo™ Luciferase Assay System and the Chroma-Luc Vectors provide a method to generate red and green (dual-color) luminescence from a single sample upon a single-reagent addition.

  • Detects red and green luminescence from a single sample
  • Dual-color luciferase detection enables assay normalization, increasing accuracy
  • Simple method; reagent is added directly to cells in medium
  • Validated automated methods available

'1U.S. Patents 5,641,641, 5,650,289 and 5,814,471, Australian Patents 649289 and other patents. 2U.S. Patents 6,387,675 and 6,552,179, Australian Patent 698424 and other patents pending. 3The method of recombina

Specifications

Content And Storage -30°C to -10°C
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.