Learn More
DWK Life Sciences Kimble™ CHROMAFLEX™ Replacement Parts: End Fitting
SDP
Catalog No. 12141401
Click to view available options
Type:
Chromatography Column
End Fitting
For Use With (Equipment):
420836-0000, 420876-0000
420836-0020, 420876-0020
420836-1620, 420876-1620
Replacement end fitting with bed support for Kimble™ CHROMAFLEX™ Flow Adapters
1.0 cm End Fitting with Bed Support, 1/16 in, for 420836-0000, 420876-0000
TRUSTED_SUSTAINABILITY
Specifications
Type Chromatography Column
Quantity 1
For Use With (Equipment) 420836-0000, 420876-0000
For Use With (Application) for 1/16 in. tubing
Includes Bed Support
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.