Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Chroman 1

Catalog No. 716310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-631-0 10 mg
71-635-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-631-0 Supplier Tocris Bioscience™ Supplier No. 7163/10
Only null left
Add to Cart
Add to Cart

Highly potent and selective ROCK 2 inhibitor; improves cell survival after cryogeneisis

Chroman 1 is a highly potent and selective ROCK2 (Rho-kinase) inhibitor (IC50 values are 1 and 52 pM at ROCK2 and ROCK1, respectively). Chroman 1 exhibits >2000-fold selectivity for ROCK2 over a range of related kinases including MRCK, PKA, and AKT1 (IC50 values are 150, >20000, and >20000 nM, respectively). Chroman 1 promotes survival of pluripotent stem cells (PSCs) in culture when used in combination with Emricasan (Cat. No. 7310), Polyamines, and Trans-ISRIB (Cat. No. 5284), a combination known as CEPT. When used as part of the CEPT cocktail, Chroman 1 also improves survival of differentiated cells following cryopreservation with cardiomyocyte cell survival increasing by 36% and motor neuron survival improving by 63% compared with DMSO controls.

Specifications

Chemical Name or Material (3 S)-N-[2-[2-(Dimethylamino)ethoxy]-4-(1 H-pyrazol-4-yl)phenyl]-3,4-dihydro-6-methoxy-2 H-1-benzopyran-3-carboxamide dihydrochloride
Quantity 10 mg
Target Rho-kinase Inhibitors
Molecular Formula C24H28N4O40.2 HCl
Formula Weight 3081
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.