Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromatin-Modifying Protein 2a Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABS1680MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1680MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1680MI Supplier MilliporeSigma Supplier No. ABS1680

Rabbit Polyclonal Antibody

Anti-Chromatin-Modifying Protein 2a is a rabbit polyclonal antibody that targets Charged multivesicular body protein 2a and has been tesed for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chromatin-Modifying Protein 2a
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CHMP2A,BC2,CHMP2
Host Species Rabbit
Immunogen GST-tagged full-length human recombinant Chromatin-modifying protein 2a.
Purification Method Affinity Purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_055268
Test Specificity This polyclonal antibody specifically detects human Chromatin-modifying protein 2a.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.