Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromatin-Modifying Protein 6 Rabbit anti-Human, Monkey, Polyclonal, MilliporeSigma™

Catalog No. ABS1645MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1645MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1645MI Supplier MilliporeSigma Supplier No. ABS1645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Chromatin-Modifying Protein 6 is a rabbit polyconal antibody that is used to detect Chromatin-Modifying Protein 6 and has been tested for use in Immunofluorescence and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chromatin-Modifying Protein 6
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CHMP6,VPS20
Host Species Rabbit
Immunogen GST-tagged full length recombinant human Chromatin-modifying protein 6.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_078867
Test Specificity This polyclonal antibody detects chromatin-modifying protein 6 (CHMP6) in human and monkey cells.
Target Species Human, Monkey
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.