Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ahlstrom-Munksjo Chromatography, Electrophoresis and Blotting Paper, Grade 238

Catalog No. 22279539 Shop All Ahlstrom Products
Change view
Click to view available options
Dimensions (L x W):
70 x 100 mm
200 x 200 mm
330 x 430 mm
460 x 570 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-279539 460 x 570 mm
09-790-53H 330 x 430 mm
09-790-53B 70 x 100 mm
09-790-53G 200 x 200 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-279539 Supplier Ahlstrom-Munksjo Supplier No. 23884657
Add to Cart
Add to Cart

Medium thick paper is recommended for general chromatography application. Also ideal for use as wicks in protein or nucleic acid blotting to provide a uniform flow or buffer through the gel to transfer membrane in a blotting sandwich.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Chromatography applications. Use as wicks in protein or nucleic acid blotting. Suited for electrophoresis
Grade 238
Thickness (Metric) 0.35 mm
Dimensions (L x W) 460 x 570 mm
Length (Metric) 460 mm
Width (Metric) 570 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.