Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Chromium
SCI_ED

Catalog No. s25249a Shop All Fisherbrand Products
Click to view available options
:
100g

Specifications

Color Silver to Gray
CAS 7440-47-3
Density 7.2g/mL at 28°C
Molecular Formula Cr
Linear Formula Cr
MDL Number MFCD00010944
Quantity 100 g
Synonym chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES [Cr]
IUPAC Name chromium
Molecular Weight (g/mol) 52.00
PubChem CID 23976
ChEBI CHEBI:28073
Formula Weight 52
Packaging Solid
Chemical Name or Material Chromium
Show More Show Less
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.