Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium carbide, 99.5% (metals basis)

Catalog No. AA1154718
Click to view available options
Quantity:
50 g
250 g

Catalog No. AA1154718

Supplier: Thermo Scientific Chemicals 01154718

Only null left
Add to Cart

CAS: 12012-35-0 | C2Cr3 | 180.01 g/mol

Chromium carbide is useful in the surface treatment of metal components. It is used to coat the surface of another metal in a technique known as thermal spraying. It is also used as an additive in cutting tools made out of cemented carbides, in order to improve toughness by preventing the growth of large grains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12012-35-0
Molecular Formula C2Cr3
Molecular Weight (g/mol) 180.01
MDL Number MFCD00016041
InChI Key UFGZSIPAQKLCGR-UHFFFAOYSA-N
Synonym chromium carbide, chromic carbide, chromium carbide,-325 mesh, amperit? 580.002, chromium 2+ ;methanidylidynechromium
PubChem CID 3650773
IUPAC Name chromium(2+);methanidylidynechromium
SMILES [Cr]#C[Cr]C#[Cr]

Specifications

Melting Point 1890°C
Physical Form Powder
Quantity 50 g
UN Number UN3178
Solubility Information Insoluble in water.
Formula Weight 180.02
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Chromium carbide
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber 234-576-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.