Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium carbide, 99.5% (metals basis)

Catalog No. AA4231318
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 12105-81-6 | Cr23C6

Chromium carbide is useful in the surface treatment of metal components. Chromium carbide is used to coat the surface of another metal in a technique known as thermal spraying. Chromium carbide is used as an additive in cutting tools made out of cemented carbides, in order to improve toughness by preventing the growth of large grains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12105-81-6
Molecular Formula Cr23C6
MDL Number MFCD00678358
Synonym bis methane trichromium

Specifications

Color Gray
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.5% (Metal basis)
Solubility Information Reacts with water.
Formula Weight 1267.97
Odor Odorless
Appearance Gray powder
Chemical Name or Material Chromium carbide
Hazard Category H228
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation DOT Class: 4.1, Packaging Group: III
EINECSNumber 235-160-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.