Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium etchant

Catalog No. p-4408703
Click to view available options
Quantity:
1 qt.
1 gal.

High Purity Ceric Ammonium Nitrate System For Precise, Clean Etching of Chromium and Chromium Oxide Films

High purity ceric ammonium nitrate system for precise, clean etching of chromium and chromium oxide films

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Chemical Name or Material Chromium etchant
Density 1.16 g/cm3 at 20°C
Quantity 1 gal.
Odor Acrid
Vapor Pressure 17mm Hg at 20°C
Form Liquid
MDL Number MFCD07370678

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.