Alfa Aesar™ Chromium(II) chloride, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 4717614

Quantity 25g

 View more versions of this product

Catalog No. AA4717614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10049-05-5
    Linear Formula CrCl2
    Formula Weight 122.9
    MDL Number MFCD00010947
    UN Number UN3260
    Merck Index 14,2244
    Synonym Chromous chloride
    Chemical Name or Material Chromium(II) chloride

    Specifications

    Formula Weight 122.9
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    TSCA TSCA
    Odor Odorless
    UN Number UN3260
    Color Gray
    Melting Point 824°C
    Quantity 25g
    Health Hazard 1 Danger
    Density 2.878g/mL
    DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    Mesh Size −80 Mesh
    Sensitivity Air & Moisture Sensitive
    CAS 10049-05-5
    RTECS Number GB5250000
    Linear Formula CrCl2
    MDL Number MFCD00010947
    Merck Index 14,2244
    Synonym Chromous chloride
    Physical Form Powder
    Chemical Name or Material Chromium(II) chloride
    Assay 99.9%, (metals basis)

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit