Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) 2,4-pentanedionate, 97%

Catalog No. p-7049030
Click to view available options
Quantity:
100 g
500 g

Catalyst in the oxidation of methacrylic esters by hydrogen peroxide, MOCVD | CAS: 21679-31-2 | C15H21CrO6 | 349.32 g/mol

Catalyst in the oxidation of methacrylic esters by hydrogen peroxide, MOCVDChromium(III) 2,4-pentanedionate is used in the reduction of detonation of nitromethane. It is also used in NMR spectroscopy as a relaxation agent due to its solubility in nonpolar organic solvents and its paramagnetism. Further, it is involved in the modification of the surface properties of solid polyurethanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21679-31-2
Molecular Formula C15H21CrO6
Molecular Weight (g/mol) 349.32
MDL Number MFCD00000015 MFCD00000015
InChI Key JWORPXLMBPOPPU-LNTINUHCSA-K
Synonym chromium iii acetylacetonate, dsstox_cid_30421, dsstox_gsid_51863
PubChem CID 91759531
IUPAC Name chromium;(Z)-4-oxoniumylidenepent-2-en-2-olate
SMILES [Cr+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 216°C
Density 1.35 g/mL
Boiling Point 340°C
Quantity 100 g
Solubility Information Soluble in toluene,acetic acid and non-polar organic solvents; Insoluble in water.
Formula Weight 349.33
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Chromium(III) 2,4-pentanedionate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 244-526-0
RTECSNumber GB7575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.