Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Chromium(III) Chloride Hexahydrate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Description
Chromium(III) chloride (CrCl3. 6H2O) hexahydrate can be used:As a source of chromium(II), which is in turn used as a catalyst in iodovinylation of aldehydes (Takai reaction).In the synthesis of chromium(III) complexes of tetradentate Schiff bases.In the preparation of Cr(III)-fatty acid complexes which can further be used as a catalyst for the synthesis of 1-monoacyIgIyceroIs.In combination with choline chloride as a deep eutectic solvent, which can be utilized as a catalyst for the esterification of formic and acetic acid with alcohols.
Specifications
Specifications
| CAS | 10060-12-5 |
| Density | 1.76 g/mL at 25°C |
| Molecular Formula | Cl3Cr -+ 6H2O |
| Linear Formula | CrCl3 -+ 6H2O |
| MDL Number | MFCD00149660 |
| Quantity | 100 g |
| Synonym | Chromium trichloride hexahydrate; Hexaaquachromium(III) chloride |
| Molecular Weight (g/mol) | 266.45 g/mol |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction