Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromium(III) chloride hexahydrate, 98%
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Melting Point | 83.0°C |
| Color | Green |
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| Assay Percent Range | 98% |
| Linear Formula | [Cr(H2O)4Cl2]Cl·2H2O |
| Merck Index | 14, 2222 |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol, slightly soluble in acetone, practically insoluble in ether |
| Formula Weight | 266.44 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. May cause an allergic skin reaction. May be corrosive to metals. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Absorb spillage to prevent material damage. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing. IF ON SK |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction