Learn More
Chromium(III) chloride, anhydrous, 99.9% (metals basis)
Description
The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties. It is used as a catalyst, for organic synthesis, for manufacturing dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | ∽1150°C |
Color | Violet |
Physical Form | Flakes |
Quantity | 25 g |
Assay Percent Range | 99.9% (Metals basis) |
Solubility Information | Soluble in alcohol,water. |
Formula Weight | 158.35 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 2.87 |
Show More |
Safety and Handling
Hazard Category | H290-H302-H317 |
Precautionary Statement | P234-P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P390-P501c |
DOTInformation | DOT Class: 8, Packaging Group: III |
EINECSNumber | 233-038-3 |
RTECSNumber | GB5425000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.