Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) fluoride, 98%

Catalog No. p-7050160
Click to view available options
Quantity:
2 g
10 g

CAS: 7788-97-8

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7788-97-8
MDL Number MFCD00016045
Synonym chromium fluoride, chromic fluoride, chromium 3+ trifluoride, acmc-1bht2, chromium 3+ ion trifluoride, chromic fluoride, solid un1756 corrosive, chromic fluoride, solution un1757 corrosive

Specifications

Melting Point >1000°C
Color Green
Physical Form Powder
Quantity 2 g
UN Number UN1756
Merck Index 14,2223
Formula Weight 108.99
Percent Purity 99.98%
Odor Odorless
Sensitivity Hygroscopic
Density 3.8 g/mL
Assay (metals basis)
Chemical Name or Material Chromium(III) fluoride
Show More Show Less
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHROMIC FLUORIDE, SOLID
EINECSNumber 232-137-9
RTECSNumber GB6125000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.