Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromium(III) fluoride hydrate, Thermo Scientific™
Click to view available options
Quantity:
1 kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Melting Point | >1000°C |
| Color | Green |
| Physical Form | Crystalline |
| Quantity | 1 kg |
| UN Number | UN1756 |
| Merck Index | 14,2223 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 108.99 (Anhydrous) |
| Sensitivity | Hygroscopic |
| Chemical Name or Material | Chromium(III) fluoride hydrate |
Safety and Handling
| Hazard Category | H302+H312+H332-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H302-H312-H332 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHROMIC FLUORIDE, SOLID |
| EINECSNumber | 232-137-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction