Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) oxide, 99.6% (metals basis)

Catalog No. aa3625814
Click to view available options
Quantity:
25 g
100 g

CAS: 1308-38-9 | Cr2O3 | 151.99 g/mol

For vacuum deposition. It can be used in selective oxidation. It is mainly used for smelting metal chromium and chromium carbide. It is used as coloring agent for color painting on enamelware and porcelain ware, imitation leather or building material. It is also used for making sun-proof coating, abrasive, green polishing compound and special printing ink for printing bank note. It is used as catalyst for organic synthetic chromium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-38-9
Molecular Formula Cr2O3
Molecular Weight (g/mol) 151.99
MDL Number MFCD00010949
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
Synonym chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
PubChem CID 517277
ChEBI CHEBI:48242
IUPAC Name dichromium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]

Specifications

Physical Form Fused Lump
Quantity 25 g
Assay Percent Range 99.6% (metals basis)
Formula Weight 151.99
Density 5.22g/mL
Chemical Name or Material Chromium oxide
EINECSNumber 215-160-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.