Alfa Aesar™ Chromium(III) potassium sulfate dodecahydrate, 98%

Manufacturer: Alfa Aesar™ A1181636

Quantity 500g

 View more versions of this product

Catalog No. AAA1181636

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7788-99-0
    Molecular Formula CrKO8S2·12H2O
    Linear Formula CrK(SO4)2·12H2O
    Formula Weight 499.39 (283.21 Anhydrous)
    MDL Number MFCD00149917
    Merck Index 14,2229
    Synonym Chrome alum
    Chemical Name or Material Chromium (III) potassium sulfate dodecahydrate

    Specifications

    Formula Weight 499.39 (283.21 Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    TSCA TSCA
    MDL Number MFCD00149917
    CAS 7788-99-0
    Melting Point 89°C
    Health Hazard 1 Warning
    RTECS Number GB6850000
    Molecular Formula CrKO8S2·12H2O
    Merck Index 14,2229
    Synonym Chrome alum
    Quantity 500g
    Linear Formula CrK(SO4)2·12H2O
    Chemical Name or Material Chromium (III) potassium sulfate dodecahydrate
    Assay 98%

    Description

    Applications
    Chromium(III) potassium sulfate is used for tanning of leather and as a photographic film hardener. It is used in manufacturing of chromium-containing catalyst.It is used in mordantin in printing and dyeing industry and it could also be used as analytical reagent. In addition, it is also used as food additive.

    Solubility
    Soluble in water. 24 g/100 mL (15°C)

    Notes
    Incompatible with magnesium, aluminium. Store away from heat.

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit