Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) potassium sulfate dodecahydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number AA4408314
Click to view available options
Quantity:
25 g
100 g
500 g
7788-99-0
CrH24KO20S2
499.386
MFCD00149917
ZFVHBEKVAITXHW-UHFFFAOYSA-J
chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
24596
potassium;chromium(3+);disulfate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]
7788-99-0
CrH24KO20S2
499.386
MFCD00149917
ZFVHBEKVAITXHW-UHFFFAOYSA-J
chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
24596
potassium;chromium(3+);disulfate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]

It is used for tanning of leather and as a photographic film hardener. It is also used in textile dye, photography, ceramics, and the preparation of chromium standard solutions, as a catalyst, mordant, ceramics, tannage and developer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 89°C
Color Violet
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Soluble in water 24g/100 mL (15°C).
Formula Weight 499.39 (283.21 Anhydrous)
Odor Odorless
Appearance Violet crystals
Density 1.83 g/cm3
Chemical Name or Material Chromium(III) potassium sulfate dodecahydrate
Show More Show Less
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 233-401-6
RTECSNumber GB6850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit