Learn More
Description
This product is for educational use - no C of A is available.
Specifications
Specifications
| Color | Purple |
| Quantity | 100 g |
| Linear Formula | Cr(NO3)3·9H2O |
| Formula Weight | 400.18 |
| Grade | Reagent |
| Packaging | Reagent Grade |
| Density | 1.8g/mL |
| Chemical Name or Material | Chromium Nitrate, Nonahydrate |
Safety and Handling
| ShelfLife | 1 year |
| DOTInformation | DOT Class 5.1, III, : Oxidizer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
