Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium selenide, Black powder, 99.5% (Metals basis), Thermo Scientific Chemicals

Catalog No. AA1311914
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1311914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1311914 Supplier Thermo Scientific Chemicals Supplier No. 01311914
Only null left
Add to Cart

It is used in semiconductor, chemical vapor deposition (CVD) and physical vapor deposition (PVD) display and optical applications. It is used for coating solar energy or fuel cells and flip-chip applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12053-13-3
Molecular Formula CrSe
Molecular Weight (g/mol) 130.967
MDL Number MFCD00049885
InChI Key UVZCKRKEVWSRGT-UHFFFAOYSA-N
Synonym chromium selenide, chromium ii selenide
PubChem CID 82886
IUPAC Name selanylidenechromium
SMILES [Cr]=[Se]

Specifications

Color Black
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.5% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 130.96
Odor Odorless
Chemical Name or Material Chromium selenide
Hazard Category H301+H331-H373
Hazard Statement When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 234-999-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.