Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromium Trioxide, Technical Flake (Chromic Acid), Reagents
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
50 kg
Specifications
Specifications
Boiling Point | 250°C |
Melting Point | 196°C |
Color | Dark Red-Black |
Physical Form | Solid |
Quantity | 25 kg |
Grade | Technical |
Packaging | Steel Drum |
Specific Gravity | 2.7 |
Chemical Name or Material | Chromium Trioxide, Technical Flake (Chromic Acid) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction