Alfa Aesar™ Chromium(VI) oxide, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 1069114

Quantity 25g

 View more versions of this product

Catalog No. AA1069114

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 1333-82-0
    Formula Weight 99.99
    MDL Number MFCD00010952
    UN Number UN1463
    Merck Index 14,2235
    Chemical Name or Material Chromium(VI) oxide

    Specifications

    Formula Weight 99.99
    Health Hazard 3 GHS P Statement
    P221-P283-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see ... on this label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H271-H301-H310-H330-H334-H340-H350-H372-H361-H314-H318-H317
    May cause fire or explosion
    strong oxidizer.
    Toxic if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause genetic defects.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of damaging fertility or the unborn child.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    UN Number UN1463
    TSCA TSCA
    Sensitivity Hygroscopic
    MDL Number MFCD00010952
    Melting Point 196°C
    Quantity 25g
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: CHROMIUM TRIOXIDE, ANHYDROUS
    Physical Form Crystalline
    CAS 1333-82-0
    Boiling Point 250°C (dec.)
    Merck Index 14,2235
    Color Dark red
    Density 2.7g/mL
    RTECS Number GB6650000
    Health Hazard 1 Danger
    Chemical Name or Material Chromium(VI) oxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H271-H301-H310-H330-H334-H340-H350-H372-H361-H314-H318-H317
    May cause fire or explosion
    strong oxidizer.
    Toxic if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause genetic defects.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of damaging fertility or the unborn child.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P221-P283-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see ... on this label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit