Learn More
Description
Host Species: Mouse
Clone: LK2H10
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Pig, Rat and Rabbit. Others not known
Immunogen: Human pheochromocytoma
Molecular Weight: 68-75 kDa
Positive Control: PC-12 cells, Pancreas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
Specifications
| Antigen | Chromogranin A Ab-1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | LK2H10 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | Human pheochromocytoma |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.