Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromogranin A Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS324P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS324P 1 mL Unconjugated Concentrated 200μg/mL
MS324P1 500 μL Unconjugated Concentrated 200μg/mL
MS324B0 100 μL Biotin Concentrated 200μg/mL
MS324B1 500 μL Biotin Concentrated 200μg/mL
MS324B 1 mL Biotin Concentrated 200μg/mL
MS324P0 100 μL Unconjugated Concentrated 200μg/mL
MS324P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS324R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS324P Supplier Epredia Supplier No. MS324P
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Chromogranin A Ab-1, Mouse Monoclonal Antibody.

Chromogranin A (a protein of 439-amino acids which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.

Host Species: Mouse

Clone: LK2H10

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Pig, Rat and Rabbit. Others not known

Immunogen: Human pheochromocytoma

Molecular Weight: 68-75 kDa

Positive Control: PC-12 cells, Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Specifications

Antigen Chromogranin A Ab-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone LK2H10
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human pheochromocytoma
Quantity 1 mL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human, Non-human Primate, Pig, Rabbit, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.