Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Chromogranin A Ab-3, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Chromogranin A Ab-3, Mouse Monoclonal Antibody.

$151.00 - $520.50

Specifications

Antigen Chromogranin A Ab-3
Clone LK2H10 + PHE5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS382P
View Documents
Thermo Scientific™
MS382P
Purified Ab with BSA and Azide 1mL
Each for $520.50
Add to cart
 
MS382P0
View Documents
Thermo Scientific™
MS382P0
Purified Ab with BSA and Azide 100μL
Each for $151.00
Add to cart
 
MS382P1
View Documents
Thermo Scientific™
MS382P1
Purified Ab with BSA and Azide 500μL
Each for $300.50
Add to cart
 
MS382P1ABX
View Documents
Thermo Scientific™
MS382P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $377.50
Add to cart
 
MS382PABX
View Documents
Thermo Scientific™
MS382PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $477.50
Add to cart
 
MS382R7
View Documents
Thermo Scientific™
MS382R7
Ready-to-use for IHC (UVLP) 7mL
Each for $296.50
Add to cart
 
MS382R1
View Documents
Thermo Scientific™
MS382R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Chromogranin A Ab-3, Mouse Monoclonal Antibody.

Chromogranin A (a protein of 439-amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Ab-3 cocktail is especially designed for sensitive detection of Chromogranin A in formalin-fixed, paraffin-embedded tissues.

Host Species: Mouse

Clone: LK2H10 + PHE5

Isotype: IgG1 + IgG1

Species Reactivity: Human and Monkey. Others not known.

Immunogen: Human pheochromocytoma

Molecular Weight: 68-75 kDa

Positive Control: Pancreas

Cellular Localization: Cytoplasmic in endocrine cells

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Chromogranin A Ab-3
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Primary
IgG1
LK2H10 + PHE5
Monoclonal
Murine
Oncogenesis
Human pheochromocytoma
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit