Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromogranin A Antibody - BSA Free, Novus Biologicals™

Sheep Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Chromogranin A |
---|---|
Concentration | 5 mg/mL |
Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:25-1:100 |
Applications | ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017479
![]() |
Novus Biologicals
NB100624640.1ML |
0.1 mL |
Each for $206.50
|
|
|||||
NB10062464
![]() |
Novus Biologicals
NB10062464 |
0.5 mL |
Each for $499.50
|
|
|||||
Description
Chromogranin A Polyclonal specifically detects Chromogranin A in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Chromogranin A | |
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:25-1:100 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
P10645 | |
1113 | |
Synthetic peptide corresponding to the amino acids 1 - 15 of the native molecule conjugated to Keyhole limpet Haemocyanin. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
5 mg/mL | |
ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry | |
Unconjugated | |
Sheep | |
Cancer, Neuronal Cell Markers | |
PBS with 0.09% Sodium Azide | |
betagranin (N-terminal fragment of chromogranin A), CGA, chromogranin A (parathyroid secretory protein 1), chromogranin-A, parathyroid secretory protein 1, Pituitary secretory protein I, SP-I | |
CHGA | |
IgG | |
IgG purified | |
This product recognizes human chromogranin A, a secreted peptide that inhibits glucose-induced insulin release. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title