Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromogranin A Antibody (CHGA/1815R), Novus Biologicals™

Rabbit Monoclonal Antibody
$278.52 - $554.39
Specifications
| Antigen | Chromogranin A |
|---|---|
| Clone | CHGA/1815R |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25320220
![]() |
Novus Biologicals
NBP25320220UG |
20 μg |
Each for $278.52
|
|
|||||
NB253202100
![]() |
Novus Biologicals
NBP253202100UG |
100 μg |
Each for $554.39
|
|
|||||
NB253202200
![]() |
Novus Biologicals
NBP253202200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Chromogranin A Monoclonal specifically detects Chromogranin A in Human, Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Chromogranin A | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Neuronal Cell Markers, Tumor Biomarkers | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 1113 | |
| Recombinant full-length human CHGA protein | |
| Primary | |
| Store at 4C. |
| CHGA/1815R | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat (Negative) | |
| betagranin (N-terminal fragment of chromogranin A), CGA, chromogranin A (parathyroid secretory protein 1), chromogranin-A, parathyroid secretory protein 1, Pituitary secretory protein I, SP-I | |
| CHGA | |
| IgG κ | |
| Protein A or G purified | |
| 72 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title