Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromogranin A Antibody (SPM585), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234815MFV500
Description
Chromogranin A Monoclonal antibody specifically detects Chromogranin A in Human, Monkey, Mouse (Negative), Rat (Negative) samples. It is validated for Flow CytometrySpecifications
Chromogranin A | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Monkey, Mouse (Negative), Rat (Negative) | |
Purified |
Flow Cytometry | |
SPM585 | |
Flow Cytometry | |
betagranin (N-terminal fragment of chromogranin A), CGA, chromogranin A (parathyroid secretory protein 1), chromogranin-A, parathyroid secretory protein 1, Pituitary secretory protein I, SP-I | |
Human pheochromocytoma (Uniprot: P10645) | |
0.1 mL | |
Apoptosis, Cancer, Neuronal Cell Markers, Tumor Biomarkers | |
1113 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction