Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromogranin B Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297350MFV500
Description
Chromogranin B Polyclonal antibody specifically detects Chromogranin B in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Chromogranin B | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
CgB, chromogranin B (secretogranin 1), Chromogranin-B, SCG1sgI, secretogranin B, Secretogranin I, secretogranin-1, SgI | |
Produced in rabbits immunized with E. coli-derived Human Chromogranin B fragment. | |
0.1 mL | |
Cell Biology, Cytokines and Growth Factors, Neuroscience, Signal Transduction | |
1114 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction