Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromogranin B Antibody (SR1105), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321794100UL
Description
Chromogranin B Monoclonal antibody specifically detects Chromogranin B in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Chromogranin B | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1105 | |
Western Blot 1:500-1:2000 | |
CgB, chromogranin B (secretogranin 1), Chromogranin-B, SCG1sgI, secretogranin B, Secretogranin I, secretogranin-1, SgI | |
A synthesized peptide derived from human Chromogranin B (Uniprot #: P05060) | |
100 μg | |
Cell Biology, Cytokines and Growth Factors, Neuroscience, Signal Transduction | |
1114 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction