Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chromogranin B Monoclonal Antibody (CL11404)
GREENER_CHOICE

Catalog No. PIMA544173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544173 Supplier Invitrogen™ Supplier No. MA544173
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The antibody solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: KQYSSHHTAE KRKRLGELFN PYYDPLQWKS SHFERRDNMN DNFLEGEEEN ELTLNEKNFF PEYNYDWWEK KPFSEDVNWG YEKRNLARVP KLDLKRQYDR VAQLDQLLHY RKKSAEFPDF YDS Highest antigen sequence indentity to the following orthologs: Mouse - 73%, Rat - 73%.

Chromogranin B is a tyrosine sulfated secretory protein found in a wide variety of peptidergic endocrine cells. Chromogranin functions as a neuroendocrine secretory granule protein which may be the precursor for other biologically active peptides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chromogranin B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone CL11404
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Chgb
Gene Accession No. P05060
Gene Alias CCB peptide; CCB peptide long form; CCB peptide short form; CgB; CHGB; chromogranin B; chromogranin B (secretogranin 1); Chromogranin B, parathyroid secretory protein; chromogranin-B; GAWK peptide; Glucagonoma peptide; PE-11; SCG1; Scg-1; secretogranin B; secretogranin I; Secretogranin-1; secretogranin-I; SgI
Gene Symbols Chgb
Host Species Mouse
Immunogen Recombinant protein fragment of Human CHGB (Product #RP-89888)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1114
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.