Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chromogranin B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552954
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552954 Supplier Invitrogen™ Supplier No. PA552954
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEVDKRRTRP RHHHGRSRPD RSSQGGSLPS EEKGHPQEES EESNVSMASL GEKRDHHSTH YRASEEEPEY GEEIKGYPGV QAPEDLEWER YRGRGSEEYR APRPQSEESW DEEDKRNYPS LELDKMAHGY GEESE Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 57%.

Chromogranin B is a tyrosine sulfated secretory protein found in a wide variety of peptidergic endocrine cells. Chromogranin functions as a neuroendocrine secretory granule protein which may be the precursor for other biologically active peptides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chromogranin B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Chgb
Gene Accession No. P05060
Gene Alias CCB peptide; CCB peptide long form; CCB peptide short form; CgB; CHGB; chromogranin B; chromogranin B (secretogranin 1); Chromogranin B, parathyroid secretory protein; chromogranin-B; GAWK peptide; Glucagonoma peptide; PE-11; SCG1; Scg-1; secretogranin B; secretogranin I; Secretogranin-1; secretogranin-I; SgI
Gene Symbols Chgb
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Chromogranin B. Recombinant protein control fragment (Product #RP-89889).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1114
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.