Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromogranin C Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299416PCP
Description
Chromogranin C Polyclonal antibody specifically detects Chromogranin C in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Chromogranin C | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
CHCG, Chromogranin-C, EM66, SCG2, secretogranin IICHGCchromogranin-C, secretoneurin, SgIIsecretogranin-2, SN | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Chromogranin C. | |
0.1 mL | |
Neuroscience | |
7857 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction