Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromogranin C Antibody (SGII 6B1/3), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP276658MFV500
Description
Chromogranin C Monoclonal antibody specifically detects Chromogranin C in Rat samples. It is validated for Flow CytometrySpecifications
Chromogranin C | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
Purified |
Flow Cytometry | |
SGII 6B1/3 | |
Flow Cytometry | |
CHCG, Chromogranin-C, EM66, SCG2, secretogranin IICHGCchromogranin-C, secretoneurin, SgIIsecretogranin-2, SN | |
Peptide from rat Chromogranin C (C terminus of p18: KEENSRENPF) | |
0.1 mL | |
Neuroscience | |
7857 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction