Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromogranin C (SCG2) Monoclonal Antibody (OTI3F12), OriGene
SDP

Catalog No. 502543859
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3859 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3859 Supplier OriGene Supplier No. CF811982
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chromogranin C (SCG2) Monoclonal antibody specifically detects Chromogranin C (SCG2) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Chromogranin C (SCG2)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3F12
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene TIMM29
Gene Accession No. Q9BSF4
Gene Alias Chcg; Chgc; Chromogranin C (Secretogranin II); chromogranin-C; EM66; Manserin; SCG2; Scg-2; secretogranin 2; secretogranin II; secretogranin II (chromogranin C); secretogranin-2; Secretoneurin; SgII; SN
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 100-260 of human C19orf52 (NP_612367) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90580
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.