Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromotrope FB

Catalog No. p-7026364
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 3567-69-9 | C20H12N2Na2O7S2 | 502.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3567-69-9
Molecular Formula C20H12N2Na2O7S2
Molecular Weight (g/mol) 502.42
MDL Number MFCD00003978
InChI Key OJNIRNOPCQQNRY-JZFXEGLSSA-L
Synonym Acid Red 14; C.I. 14720
PubChem CID 44135716
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=C\C(=N/NC2=C3C=CC=CC3=C(C=C2)S([O-])(=O)=O)C(=O)C2=CC=CC=C12

Specifications

IUPAC Name disodium;4-oxo-3-[(4-sulfonatonaphthalen-1-yl)hydrazinylidene]naphthalene-1-sulfonate
Formula Weight 502.42
Quantity 10 g
Chemical Name or Material Chromotrope FB
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 222-657-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.